Gene Transcripts
- Gene Symbol
- RIN2
- Entrez Gene ID
- 54453
- Gene Name
- Ras and Rab interactor 2
- Gene Aliases
- MACS, RASSF4
- Chromosome Location
- Chr. 20: 19757610 - 20002459 on Build GRCh38
- UniGene ID
- Hs.472270
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC090059.1 |
- |
2 |
214 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000412571.1 |
- |
3 |
346 |
|
Chr. 20 - 19758148 - 19809282 [+] |
View Full Product Details