Gene Transcripts
- Gene Symbol
- ERVFC1-1
- Entrez Gene ID
- 346547
- Gene Name
- endogenous retrovirus group FC1 member 1
- Gene Aliases
- envF(c)2
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124285.1 |
- |
1 |
157 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000416982.1 |
- |
1 |
157 |
|
Chr. 7 - 153411234 - 153413963 [-] |
View Full Product Details