Gene Transcripts
- Gene Symbol
- ARHGEF3-AS1
- Entrez Gene ID
- 100874200
- Gene Name
- ARHGEF3 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 56940040 - 56960854 on Build GRCh38
- UniGene ID
- Hs.549631
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046572.1 |
|
2 |
203 |
|
GenBank mRNA |
BX090838.1 |
- |
2 |
203 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000477246.5 |
- |
2 |
203 |
|
Chr. 3 - 56940040 - 56960854 [+] |
View Full Product Details