Gene Transcripts

Gene Symbol
STX6
Entrez Gene ID
10228
Gene Name
syntaxin 6
Gene Aliases
Chromosome Location
Chr. 1: 180949690 - 181023121 on Build GRCh38
UniGene ID
Hs.518417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK056657.1 - 2 838


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000358073.2 - 2 838 Chr. 1 - 180944039 - 180976482 [-]

View Full Product Details