Gene Transcripts
- Gene Symbol
- CFAP44-AS1
- Entrez Gene ID
- 100874029
- Gene Name
- CFAP44 antisense RNA 1
- Gene Aliases
- WDR52-AS1
- Chromosome Location
- Chr. 3: 113403998 - 113433992 on Build GRCh38
- UniGene ID
- Hs.599665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046728.1 |
|
1 |
48 |
|
GenBank mRNA |
BX108680.1 |
- |
1 |
49 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000473329.1 |
- |
1 |
55 |
|
Chr. 3 - 113403991 - 113427046 [+] |
View Full Product Details