Gene Transcripts

Gene Symbol
NADK2-AS1
Entrez Gene ID
101056700
Gene Name
NADK2 antisense RNA 1
Gene Aliases
NADKD1-AS1
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.570980
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC033402.1 - 1 50


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG771949.1 - 1 55 Chr. 5 - 36221055 - 36221901 [+]
GENCODE ENST00000501794.2 - 1 50 Chr. 5 - 36221055 - 36221902 [+]

View Full Product Details