Gene Transcripts

Gene Symbol
EXTL3-AS1
Entrez Gene ID
101929402
Gene Name
EXTL3 antisense RNA 1
Gene Aliases
C8orf50
Chromosome Location
Chr. 8: 28698403 - 28701464 on Build GRCh38
UniGene ID
Hs.667366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC043205.2 - 2 1578


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000523789.2 - 3 445 Chr. 8 - 28698212 - 28700530 [-]

View Full Product Details