Gene Transcripts
- Gene Symbol
- ROPN1L-AS1
- Entrez Gene ID
- 100505845
- Gene Name
- ROPN1L antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 10440735 - 10441851 on Build GRCh38
- UniGene ID
- Hs.599395
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY053860.1 |
- |
2 |
207 |
|
Chr. 5 - 10441058 - 10441847 [-] |
GENCODE |
ENST00000513037.1 |
- |
2 |
152 |
|
Chr. 5 - 10441290 - 10441792 [-] |
GenBankmRNA |
AA398071.1 |
- |
2 |
149 |
|
Chr. 5 - 10441290 - 10441792 [-] |
View Full Product Details