Gene Transcripts

Gene Symbol
TUG1
Entrez Gene ID
55000
Gene Name
taurine up-regulated 1 (non-protein coding)
Gene Aliases
LINC00080, NCRNA00080, TI-227H
Chromosome Location
Chr. 22: 30969211 - 30979395 on Build GRCh38
UniGene ID
Hs.554829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002323.2 3 3415
NR_110492.1 4 3471
NR_110493.1 3 3401
GenBank mRNA AK000625.1 - 3 1400
AK000749.1 - 4 1316
AK056480.1 - 3 1949
AK057695.1 - 2 1439
BC037986.1 - 3 1558


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA795543.1 - 2 317 Chr. 22 - 30973370 - 30975407 [+]
GenBankmRNA AA476695.1 - 2 345 Chr. 22 - 30973343 - 30975338 [+]
GENCODE ENST00000521091.6 - 4 1315 Chr. 22 - 30971367 - 30976063 [+]
GenBankmRNA CB044441.1 - 2 553 Chr. 22 - 30973145 - 30975333 [+]
GenBankmRNA DA834152.1 - 2 635 Chr. 22 - 30973052 - 30975455 [+]
GenBankmRNA DA096209.1 - 2 420 Chr. 22 - 30973281 - 30975408 [+]
GenBankmRNA BQ932118.1 - 2 485 Chr. 22 - 30973204 - 30975601 [+]
GenBankmRNA BX643169.1 - 2 419 Chr. 22 - 30973282 - 30975466 [+]
GenBankmRNA BE079454.1 - 2 213 Chr. 22 - 30973474 - 30975670 [+]
GenBankmRNA BE079543.1 - 2 213 Chr. 22 - 30973474 - 30975694 [+]
GenBankmRNA CB141510.1 - 2 516 Chr. 22 - 30973185 - 30975335 [+]
GenBankmRNA BP240227.1 - 2 514 Chr. 22 - 30973187 - 30975316 [+]
GenBankmRNA DB070372.1 - 2 246 Chr. 22 - 30973455 - 30975559 [+]
GenBankmRNA BE694058.1 - 2 213 Chr. 22 - 30973474 - 30975692 [+]
GENCODE ENST00000602393.1 - 2 1438 Chr. 22 - 30972263 - 30976063 [+]
GENCODE ENST00000519077.3 - 3 1546 Chr. 22 - 30971066 - 30979395 [+]
GenBankmRNA DA832964.1 - 2 422 Chr. 22 - 30973283 - 30975562 [+]
GenBankmRNA CB149465.1 - 2 462 Chr. 22 - 30973226 - 30975456 [+]
GenBankmRNA CN298753.1 - 2 348 Chr. 22 - 30973343 - 30975608 [+]
GenBankmRNA DB140531.1 - 2 464 Chr. 22 - 30973225 - 30975343 [+]
GENCODE ENST00000569384.1 - 2 232 Chr. 22 - 30973412 - 30975479 [+]
GENCODE ENST00000602971.1 - 2 786 Chr. 22 - 30972901 - 30976081 [+]
GENCODE ENST00000540687.5 - 3 1949 Chr. 22 - 30970677 - 30976063 [+]

View Full Product Details