Gene Transcripts

Gene Symbol
MIR9-3HG
Entrez Gene ID
254559
Gene Name
MIR9-3 host gene
Gene Aliases
LINC00925
Chromosome Location
Chr. 15: 89361579 - 89398487 on Build GRCh38
UniGene ID
Hs.136313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015411.1 4 479
NR_133001.1 4 1062
NR_133002.1 3 360
NR_133003.1 4 394
GenBank mRNA AK054931.1 - 4 394
AK123792.1 - 1 1246
AK125292.1 - 4 405
BX441353.2 - 3 392
DA059714.1 - 4 394
DB020558.1 - 2 234
DB067527.1 - 2 172
DB068800.1 - 2 172
DB452761.1 - 2 197
DB454083.1 - 2 172


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI825996.1 - 2 165 Chr. 15 - 89379130 - 89395344 [+]
GenBankmRNA DB097661.1 - 2 172 Chr. 15 - 89379123 - 89395193 [+]
GenBankmRNA DB027914.1 - 2 170 Chr. 15 - 89379126 - 89395172 [+]
GenBankmRNA DB075548.1 - 2 172 Chr. 15 - 89379124 - 89395119 [+]
GenBankmRNA HY004906.1 - 2 214 Chr. 15 - 89379081 - 89389475 [+]
GenBankmRNA HY110386.1 - 3 331 Chr. 15 - 89371610 - 89389423 [+]
GenBankmRNA DB061891.1 - 2 173 Chr. 15 - 89379123 - 89395144 [+]
GENCODE ENST00000538734.6 - 4 405 Chr. 15 - 89378116 - 89396240 [+]
GenBankmRNA DB036793.1 - 2 259 Chr. 15 - 89379038 - 89395091 [+]
GenBankmRNA DB071179.1 - 2 257 Chr. 15 - 89379039 - 89395043 [+]
GenBankmRNA DB021481.1 - 2 257 Chr. 15 - 89379039 - 89395082 [+]
GenBankmRNA DB068534.1 - 2 85 Chr. 15 - 89378106 - 89395219 [+]
GENCODE ENST00000546186.2 - 1 1246 Chr. 15 - 89386596 - 89396240 [+]
GenBankmRNA HY056341.1 - 2 241 Chr. 15 - 89379054 - 89389426 [+]
GenBankmRNA DB066190.1 - 2 81 Chr. 15 - 89378110 - 89395222 [+]
GenBankmRNA BG819916.1 - 1 25 Chr. 15 - 89387817 - 89395478 [+]
GenBankmRNA DA451099.1 - 2 85 Chr. 15 - 89378106 - 89395242 [+]
GenBankmRNA DB060384.1 - 2 172 Chr. 15 - 89379123 - 89395137 [+]
GenBankmRNA DB068926.1 - 2 188 Chr. 15 - 89379107 - 89395137 [+]
GenBankmRNA DB035646.1 - 2 188 Chr. 15 - 89379107 - 89395151 [+]
GenBankmRNA DB058234.1 - 2 172 Chr. 15 - 89379123 - 89395134 [+]
GenBankmRNA DA804092.1 - 2 89 Chr. 15 - 89378102 - 89395217 [+]
GenBankmRNA DB035856.1 - 2 257 Chr. 15 - 89379038 - 89395086 [+]
GenBankmRNA DB093044.1 - 3 200 Chr. 15 - 89378100 - 89395123 [+]
GenBankmRNA DA340519.1 - 2 176 Chr. 15 - 89379119 - 89395185 [+]
GenBankmRNA DB021867.1 - 2 173 Chr. 15 - 89379124 - 89395144 [+]
GenBankmRNA DB057461.1 - 2 113 Chr. 15 - 89378078 - 89395195 [+]
GenBankmRNA DB042606.1 - 2 176 Chr. 15 - 89379119 - 89395151 [+]
GenBankmRNA DB023242.1 - 2 60 Chr. 15 - 89378131 - 89395268 [+]

View Full Product Details