Gene Transcripts
- Gene Symbol
- CACNA1C-AS2
- Entrez Gene ID
- 100874235
- Gene Name
- CACNA1C antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 2668500 - 2672220 on Build GRCh38
- UniGene ID
- Hs.667644
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046579.1 |
|
2 |
364 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000545526.1 |
- |
2 |
364 |
|
Chr. 12 - 2668500 - 2672220 [-] |
View Full Product Details