Gene Transcripts
- Gene Symbol
- PRICKLE1
- Entrez Gene ID
- 144165
- Gene Name
- prickle planar cell polarity protein 1
- Gene Aliases
- EPM1B, RILP
- Chromosome Location
- Chr. 12: 42458338 - 42589770 on Build GRCh38
- UniGene ID
- Hs.524348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042722.1 |
- |
2 |
1921 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000547824.1 |
- |
2 |
1921 |
|
Chr. 12 - 42459366 - 42466128 [+] |
View Full Product Details