Gene Transcripts
- Gene Symbol
- FAM181A-AS1
- Entrez Gene ID
- 283592
- Gene Name
- FAM181A antisense RNA 1
- Gene Aliases
- C14orf86
- Chromosome Location
- Chr. 14: 93904730 - 93926372 on Build GRCh38
- UniGene ID
- Hs.585648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027004.1 |
|
2 |
484 |
|
GenBank mRNA |
BC037859.2 |
- |
2 |
484 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000554742.1 |
- |
2 |
484 |
|
Chr. 14 - 93904730 - 93926372 [-] |
View Full Product Details