Gene Transcripts
- Gene Symbol
- LINC-ROR
- Entrez Gene ID
- 100885779
- Gene Name
- long intergenic non-protein coding RNA, regulator of reprogramming
- Gene Aliases
- ROR, lincRNA-RoR
- Chromosome Location
- Chr. 18: 57054571 - 57072119 on Build GRCh38
- UniGene ID
- Hs.742426
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_048536.1 |
|
4 |
1599 |
|
GenBank mRNA |
HQ315778.1 |
- |
4 |
1599 |
|
LC027886.1 |
- |
5 |
1214 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000553704.2 |
- |
4 |
1599 |
|
Chr. 18 - 57054582 - 57072119 [-] |
View Full Product Details