Gene Transcripts
- Gene Symbol
- LOC105370489
- Entrez Gene ID
- 105370489
- Gene Name
- uncharacterized LOC105370489
- Gene Aliases
- Chromosome Location
- Chr. 14: 50821797 - 50823509 on Build GRCh38
- UniGene ID
- Hs.569387
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135258.1 |
|
2 |
423 |
|
GenBank mRNA |
HY010266.1 |
- |
2 |
424 |
|
- Gene Symbol
- NIN
- Entrez Gene ID
- 51199
- Gene Name
- ninein
- Gene Aliases
- SCKL7
- Chromosome Location
- Chr. 14: 50719763 - 50831121 on Build GRCh38
- UniGene ID
- Hs.310429
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA742651.1 |
- |
1 |
216 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555966.1 |
- |
2 |
340 |
|
Chr. 14 - 50821880 - 50823501 [+] |
View Full Product Details