Gene Transcripts
- Gene Symbol
- AQP4-AS1
- Entrez Gene ID
- 147429
- Gene Name
- AQP4 antisense RNA 1
- Gene Aliases
- C18orf16, CHST9-AS1
- Chromosome Location
- Chr. 18: 26865308 - 26935946 on Build GRCh38
- UniGene ID
- Hs.559280
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026908.1 |
|
3 |
228 |
|
GenBank mRNA |
AK055069.1 |
- |
3 |
228 |
|
BC043377.1 |
- |
2 |
107 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI757858.1 |
- |
|
109 |
|
Chr. - - [-] |
GenBankmRNA |
DA197840.1 |
- |
3 |
228 |
|
Chr. 18 - 26865308 - 26933053 [+] |
GenBankmRNA |
DB464139.1 |
- |
2 |
145 |
|
Chr. 18 - 26655743 - 26933085 [+] |
GENCODE |
ENST00000568797.3 |
- |
3 |
228 |
|
Chr. 18 - 26865308 - 26935946 [+] |
GenBankmRNA |
DA214122.1 |
- |
4 |
359 |
|
Chr. 18 - 26655742 - 26930757 [+] |
GENCODE |
ENST00000578701.5 |
- |
2 |
107 |
|
Chr. 18 - 26865307 - 27190698 [+] |
View Full Product Details