Gene Transcripts
- Gene Symbol
- AQP4-AS1
- Entrez Gene ID
- 147429
- Gene Name
- AQP4 antisense RNA 1
- Gene Aliases
- C18orf16, CHST9-AS1
- Chromosome Location
- Chr. 18: 26865308 - 26935946 on Build GRCh38
- UniGene ID
- Hs.559280
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC043377.1 |
- |
4 |
315 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI757858.1 |
- |
|
316 |
|
Chr. - - [-] |
GENCODE |
ENST00000578701.5 |
- |
4 |
315 |
|
Chr. 18 - 26865307 - 27190698 [+] |
View Full Product Details