Gene Transcripts
- Gene Symbol
- MDP1
- Entrez Gene ID
- 145553
- Gene Name
- magnesium dependent phosphatase 1
- Gene Aliases
- FN6PASE, MDP-1
- Chromosome Location
- Chr. 14: 24213937 - 24216070 on Build GRCh38
- UniGene ID
- Hs.220963
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092821.1 |
- |
4 |
658 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000528804.1 |
- |
4 |
658 |
|
Chr. 14 - 24212739 - 24215987 [-] |
View Full Product Details