Gene Transcripts
- Gene Symbol
- PYCARD-AS1
- Entrez Gene ID
- 100652740
- Gene Name
- PYCARD antisense RNA 1
- Gene Aliases
- C16orf98, PYCARDOS
- Chromosome Location
- Chr. 16: 31201885 - 31203452 on Build GRCh38
- UniGene ID
- Hs.550659
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102400.1 |
|
2 |
1073 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000561916.2 |
- |
2 |
1073 |
|
Chr. 16 - 31201885 - 31203452 [+] |
View Full Product Details