Gene Transcripts

Gene Symbol
TTC23
Entrez Gene ID
64927
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
HCC-8
Chromosome Location
Chr. 15: 99136323 - 99251226 on Build GRCh38
UniGene ID
Hs.513195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC034816.1 - 1 1621


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000564527.1 - 1 1623 Chr. 15 - 99139317 - 99145370 [+]

View Full Product Details