Gene Transcripts
- Gene Symbol
- RBFOX3
- Entrez Gene ID
- 146713
- Gene Name
- RNA binding protein, fox-1 homolog 3
- Gene Aliases
- FOX-3, FOX3, HRNBP3, NEUN
- Chromosome Location
- Chr. 17: 79089345 - 79611101 on Build GRCh38
- UniGene ID
- Hs.135229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126788.1 |
- |
2 |
200 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000571452.1 |
- |
2 |
200 |
|
Chr. 17 - 79598032 - 79603921 [-] |
View Full Product Details