Gene Transcripts
- Gene Symbol
- HSPC102
- Entrez Gene ID
- 105372274
- Gene Name
- uncharacterized HSPC102
- Gene Aliases
- Chromosome Location
- Chr. 19: 11300777 - 11301535 on Build GRCh38
- UniGene ID
- Hs.606581
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF161365.1 |
- |
2 |
155 |
|
- Gene Symbol
- TSPAN16
- Entrez Gene ID
- 26526
- Gene Name
- tetraspanin 16
- Gene Aliases
- TM-8, TM4-B, TM4SF16
- Chromosome Location
- Chr. 19: 11296139 - 11326996 on Build GRCh38
- UniGene ID
- Hs.579784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI688554.1 |
- |
3 |
339 |
|
AW169944.1 |
- |
3 |
322 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000586356.5 |
- |
2 |
173 |
|
Chr. 19 - 11300777 - 11324441 [-] |
GENCODE |
ENST00000585801.1 |
- |
2 |
154 |
|
Chr. 19 - 11300867 - 11321323 [-] |
View Full Product Details