Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005259102.4 | XP_005259159.1 | 3 | 145 | |
XM_017027033.1 | XP_016882522.1 | 3 | 260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000601911.1 | - | 3 | 136 | Chr. 19 - 39314651 - 39320858 [-] |