Gene Transcripts

Gene Symbol
A1BG-AS1
Entrez Gene ID
503538
Gene Name
A1BG antisense RNA 1
Gene Aliases
A1BG-AS, A1BGAS, NCRNA00181
Chromosome Location
Chr. 19: 58351970 - 58355183 on Build GRCh38
UniGene ID
Hs.709582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015380.2 4 2077
GenBank mRNA AK027222.1 - 4 2077
BC006144.1 - 1 909
BC040926.1 - 3 1665


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000595302.1 - 4 2077 Chr. 19 - 58351970 - 58355183 [+]
GENCODE ENST00000594950.5 - 3 1665 Chr. 19 - 58347751 - 58355183 [+]

View Full Product Details