Gene Transcripts

Gene Symbol
MAGI1-IT1
Entrez Gene ID
151877
Gene Name
MAGI1 intronic transcript 1
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.476636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK056866.1 - 3 2406


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000460754.1 - 3 2406 Chr. 3 - 65872815 - 65954558 [-]

View Full Product Details