Gene Transcripts
- Gene Symbol
- MAGI1-IT1
- Entrez Gene ID
- 151877
- Gene Name
- MAGI1 intronic transcript 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.476636
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056866.1 |
- |
3 |
1976 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000460754.1 |
- |
3 |
1976 |
|
Chr. 3 - 65872815 - 65954558 [-] |
View Full Product Details