Gene Transcripts
- Gene Symbol
- SPANXA2-OT1
- Entrez Gene ID
- 619455
- Gene Name
- SPANXA2 overlapping transcript 1
- Gene Aliases
- CXorf18
- Chromosome Location
- Chr. X: 141502849 - 141649939 on Build GRCh38
- UniGene ID
- Hs.590784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037183.2 |
|
4 |
1126 |
|
GenBank mRNA |
AK093505.1 |
- |
4 |
1141 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000622372.1 |
- |
|
1126 |
|
Chr. - - [-] |
View Full Product Details