Gene Transcripts
- Gene Symbol
- BIRC6-AS2
- Entrez Gene ID
- 103752586
- Gene Name
- BIRC6 antisense RNA 2
- Gene Aliases
- linc-birc6, megamind
- Chromosome Location
- Chr. 2: 32557520 - 32574818 on Build GRCh38
- UniGene ID
- Hs.666092
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125793.1 |
|
2 |
371 |
|
GenBank mRNA |
BX093813.1 |
- |
2 |
371 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000623382.1 |
- |
|
371 |
|
Chr. - - [-] |
View Full Product Details