Gene Transcripts
- Gene Symbol
- Sfpq
- Entrez Gene ID
- 71514
- Gene Name
- splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
- Gene Aliases
- 1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
- Chromosome Location
- Chr. 4: 127020560 - 127037016 on Build GRCm38
- UniGene ID
- Mm.257276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141302.1 |
- |
1 |
530 |
|
BC033603.1 |
- |
1 |
1928 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV071279.1 |
- |
|
51 |
|
Chr. - - [-] |
GenBankmRNA |
BE950642.1 |
- |
1 |
88 |
|
Chr. 4 - 127033731 - 127037013 [+] |
View Full Product Details