Gene Transcripts

Gene Symbol
Mkln1os
Entrez Gene ID
100503869
Gene Name
muskelin 1, intracellular mediator containing kelch motifs, opposite strand
Gene Aliases
Gm13845, OTTMUSG00000014733
Chromosome Location
Chr. 6: 31366885 - 31398773 on Build GRCm38
UniGene ID
Mm.370532
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040300.1 1 148
GenBank mRNA AK043851.1 - 1 45
AK045552.1 - 1 136


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY726223.1 - 1 128 Chr. 6 - 31383041 - 31398753 [-]
GenBankmRNA DN174269.1 - 1 149 Chr. 6 - 31367376 - 31398773 [-]
GenBankmRNA BB644004.1 - 1 132 Chr. 6 - 31385878 - 31398757 [-]
GenBankmRNA CA464169.1 - 1 135 Chr. 6 - 31367082 - 31398757 [-]

View Full Product Details