Gene Transcripts
- Gene Symbol
- Tmem132cos
- Entrez Gene ID
- 67351
- Gene Name
- transmembrane protein 132C, opposite strand
- Gene Aliases
- 1700085B03Rik
- Chromosome Location
- Chr. 5: 127256860 - 127281623 on Build GRCm38
- UniGene ID
- Mm.320172
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU946175.1 |
- |
3 |
524 |
|
Chr. 5 - 127256861 - 127281606 [-] |
GenBankmRNA |
CA466084.1 |
- |
3 |
515 |
|
Chr. 5 - 127256824 - 127281597 [-] |
GenBankmRNA |
CN840709.1 |
- |
|
512 |
|
Chr. - - [-] |
GenBankmRNA |
BY707058.1 |
- |
3 |
501 |
|
Chr. 5 - 127256862 - 127281585 [-] |
View Full Product Details