Gene Transcripts

Gene Symbol
Dlx6os1
Entrez Gene ID
320038
Gene Name
distal-less homeobox 6, opposite strand 1
Gene Aliases
A230055N17Rik, Dlx6as1, Evf-1, Evf-2, Evf1, Evf1/2, Evf2, Shhrs, mEvf-1
Chromosome Location
Chr. 6: 6820546 - 6869533 on Build GRCm38
UniGene ID
Mm.94021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015388.1 2 2400
GenBank mRNA AK032180.1 - 2 2300
AK032537.1 - 2 2400
AK038694.1 - 2 2242
AK044034.1 - 2 2259

View Full Product Details