Gene Transcripts
- Gene Symbol
- Vmn1r48
- Entrez Gene ID
- 113845
- Gene Name
- vomeronasal 1 receptor 48
- Gene Aliases
- V1ra3
- Chromosome Location
- Chr. 6: 90035933 - 90036841 on Build GRCm38
- UniGene ID
- Mm.425336
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053218.1 |
NP_444448.1
|
1 |
382 |
|
- Gene Symbol
- Vmn1r47
- Entrez Gene ID
- 113846
- Gene Name
- vomeronasal 1 receptor 47
- Gene Aliases
- V1RA7, V1ra4
- Chromosome Location
- Chr. 6: 90021888 - 90022820 on Build GRCm38
- UniGene ID
- Mm.377143
- Species
- Mus musculus
View Full Product Details