Gene Transcripts

Gene Symbol
Vmn1r43
Entrez Gene ID
113847
Gene Name
vomeronasal 1 receptor 43
Gene Aliases
V1ra2, V1ra5
Chromosome Location
Chr. 6: 89869461 - 89870529 on Build GRCm38
UniGene ID
Mm.431989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053220.2 NP_444450.2 1 444
GenBank mRNA BC125364.1 - 1 444
BC132284.1 - 1 444 40130561

View Full Product Details