Gene Transcripts
- Gene Symbol
- Vmn1r54
- Entrez Gene ID
- 113851
- Gene Name
- vomeronasal 1 receptor 54
- Gene Aliases
- V1ra9, VN7
- Chromosome Location
- Chr. 6: 90269106 - 90270053 on Build GRCm38
- UniGene ID
- Mm.377148
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053224.1 |
NP_444454.1
|
1 |
782 |
|
View Full Product Details