Gene Transcripts
- Gene Symbol
- Psg25
- Entrez Gene ID
- 114868
- Gene Name
- pregnancy-specific glycoprotein 25
- Gene Aliases
- cea13
- Chromosome Location
- Chr. 7: 18519702 - 18532264 on Build GRCm38
- UniGene ID
- Mm.389809
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054060.1 |
NP_473401.1
|
1, 2 |
249 |
|
GenBank mRNA |
BC100352.1 |
- |
1, 2 |
249 |
|
View Full Product Details