Gene Transcripts

Gene Symbol
Psg25
Entrez Gene ID
114868
Gene Name
pregnancy-specific glycoprotein 25
Gene Aliases
cea13
Chromosome Location
Chr. 7: 18519702 - 18532264 on Build GRCm38
UniGene ID
Mm.389809
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054060.1 NP_473401.1 5 1363
GenBank mRNA BC100352.1 - 5 1363

View Full Product Details