Gene Transcripts

Gene Symbol
Alpk3
Entrez Gene ID
116904
Gene Name
alpha-kinase 3
Gene Aliases
AW319487, D330016D04, MAK, Midori, mKIAA1330
Chromosome Location
Chr. 7: 81057600 - 81105612 on Build GRCm38
UniGene ID
Mm.475768
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054085.2 NP_473426.2 13 4696
GenBank mRNA AF338872.1 - 13 4690
AK052268.1 - 3 265
AK220384.1 - 9 3172

View Full Product Details