Gene Transcripts

Gene Symbol
Olfr74
Entrez Gene ID
117005
Gene Name
olfactory receptor 74
Gene Aliases
MOR174-4, mOR-EV
Chromosome Location
Chr. 2: 87973707 - 87974663 on Build GRCm38
UniGene ID
Mm.196679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054091.2 NP_473432.2 1 257
GenBank mRNA AB061229.1 - 1 257

View Full Product Details