Gene Transcripts

Gene Symbol
CNTN3
Entrez Gene ID
5067
Gene Name
contactin 3
Gene Aliases
BIG-1, PANG, PCS
Chromosome Location
Chr. 3: 74262568 - 74614448 on Build GRCh38
UniGene ID
Hs.12723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020872.2 NP_065923.1 3 256
XM_005264757.3 XP_005264814.1 4 366
XM_011533768.2 XP_011532070.1 3 3888
XM_017006507.1 XP_016861996.1 4 936
XM_017006508.1 XP_016861997.1 3 161
GenBank mRNA BC150608.1 - 3 256
DR002512.1 - 3 255

View Full Product Details