Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080446.2 | NP_536694.2 | 12 | 3072 | |
GenBank mRNA | AB048542.1 | - | 12 | 2997 | |
AK131122.1 | - | 12 | 3094 | ||
BC038261.1 | - | 2 | 803 | ||
BC068140.1 | - | 12 | 3000 | 6843107 |