Gene Transcripts

Gene Symbol
Zcchc14
Entrez Gene ID
142682
Gene Name
zinc finger, CCHC domain containing 14
Gene Aliases
AA792890, BDG29
Chromosome Location
Chr. 8: 121598697 - 121652761 on Build GRCm38
UniGene ID
Mm.103202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080855.2 NP_543131.1 4 435
GenBank mRNA AB030244.1 - 4 446
BC132449.1 - 4 433 40130726
BC137853.1 - 4 433 8860872
CB518791.1 - 2 81
CJ102007.1 - 4 278

View Full Product Details