Gene Transcripts

Gene Symbol
Vgll1
Entrez Gene ID
170828
Gene Name
vestigial like 1 homolog (Drosophila)
Gene Aliases
Tondu
Chromosome Location
Chr. X: 57088079 - 57106539 on Build GRCm38
UniGene ID
Mm.190616
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133251.1 NP_573514.1 5 544
GenBank mRNA AB036708.1 - 5 544
AK146118.1 - 4 415
BC144928.1 - 5 492 9053476
BC145949.1 - 4 476 40131252

View Full Product Details