Gene Transcripts

Gene Symbol
Prima1
Entrez Gene ID
170952
Gene Name
proline rich membrane anchor 1
Gene Aliases
B230212M13Rik, BB120497, PRiMA
Chromosome Location
Chr. 12: 103194453 - 103242720 on Build GRCm38
UniGene ID
Mm.217019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133364.2 NP_579942.2 4 388
GenBank mRNA AK021000.1 - 1 163
AK039705.1 - 1 81
AK140034.1 - 4 389
AY043275.1 - 4 389
AY225515.1 - 4 389
BC134395.1 - 3 234
BC145454.1 - 3 232
BC171927.1 - 3 232
BC171928.1 - 3 232
BC171929.1 - 3 227

View Full Product Details