Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138665.2 | NP_619606.1 | 3 | 1138 | |
GenBank mRNA | AK140451.1 | - | 2 | 414 | |
AK159236.1 | - | 3 | 1102 | ||
AK159681.1 | - | 3 | 1102 | ||
AK159704.1 | - | 3 | 1102 | ||
AK163515.1 | - | 3 | 667 | ||
AK163660.1 | - | 2 | 414 | ||
BC003456.1 | - | 3 | 647 | 2648344 |