Gene Transcripts

Gene Symbol
Hectd1
Entrez Gene ID
207304
Gene Name
HECT domain containing 1
Gene Aliases
A630086P08Rik, AI844876, b2b327Clo, opm
Chromosome Location
Chr. 12: 51743721 - 51829605 on Build GRCm38
UniGene ID
Mm.249391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144788.2 NP_659037.2 25 4316
GenBank mRNA AK141970.1 - 3 231
AK173106.1 - 5 845

View Full Product Details