Gene Transcripts

Gene Symbol
ADCY1
Entrez Gene ID
107
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, DFNB44
Chromosome Location
Chr. 7: 45574140 - 45723116 on Build GRCh38
UniGene ID
Hs.192215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021116.2 NP_066939.1 20 3165
GenBank mRNA AB209496.1 - 20 3111
BU181704.1 - 1 98
L05500.1 - 18 2309

View Full Product Details