Gene Transcripts

Gene Symbol
Fam20a
Entrez Gene ID
208659
Gene Name
family with sequence similarity 20, member A
Gene Aliases
AI606893
Chromosome Location
Chr. 11: 109669492 - 109722510 on Build GRCm38
UniGene ID
Mm.208662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153782.1 NP_722477.1 1, 2 948
GenBank mRNA AK142932.1 - 1, 2 972
BC011274.1 - 1, 2 854
BC029169.1 - 1, 2 948 4947019

View Full Product Details