Gene Transcripts

Gene Symbol
Gps1
Entrez Gene ID
209318
Gene Name
G protein pathway suppressor 1
Gene Aliases
Cops1, Csn1, R75577, Sgn1
Chromosome Location
Chr. 11: 120784260 - 120789107 on Build GRCm38
UniGene ID
Mm.30195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177874.1 NP_001171345.1 5, 6 655
NM_145370.2 NP_663345.2 5, 6 795
GenBank mRNA AK088490.1 - 5, 6 652
BC003350.1 - 5, 6 795 3481979
BC103782.1 - 5, 6 622 6486599

View Full Product Details