Gene Transcripts

Gene Symbol
Rad51ap2
Entrez Gene ID
209550
Gene Name
RAD51 associated protein 2
Gene Aliases
EG209550
Chromosome Location
Chr. 12: 11455759 - 11517161 on Build GRCm38
UniGene ID
Mm.341257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111118.1 NP_001104588.1 1 2424

View Full Product Details